Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yellow-d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079096.3 | NP_523820.2 | 2 - 3 | 1459 | 74 | |
GenBank mRNA | BT003303.1 | - | 2 - 3 | 1499 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |