Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: enok | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299894.1 | NP_001286823.1 | 4 - 5 | 6131 | 59 | |
NM_079114.2 | NP_523838.1 | 4 - 5 | 5862 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |