Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SerT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299910.1 | NP_001286839.1 | 4 - 5 | 1668 | 88 | |
NM_001382130.1 | NP_001369117.1 | 4 - 5 | 1126 | 88 | ||
NM_079122.3 | NP_523846.2 | 4 - 5 | 1126 | 88 | ||
GenBank mRNA | AY071023.1 | - | 4 - 5 | 1123 | 88 | |
U04809.1 | - | 3 - 4 | 669 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster Inventoried |