Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274252.1 | NP_001261181.1 | 1 - 2 | 379 | 67 | |
NM_079143.3 | NP_523867.1 | 1 - 2 | 379 | 67 | ||
GenBank mRNA | BT044068.1 | - | 1 - 2 | 375 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |