Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043111.3 | NP_001036576.1 | 16 - 17 | 5798 | 70 | |
NM_001103992.2 | NP_001097462.1 | 2 - 3 | 775 | 70 | ||
NM_001274282.2 | NP_001261211.1 | 17 - 18 | 6385 | 70 | ||
NM_001274283.2 | NP_001261212.1 | 13 - 14 | 5271 | 70 | ||
NM_001274284.1 | NP_001261213.1 | 8 - 9 | 2677 | 70 | ||
NM_001299960.1 | NP_001286889.1 | 2 - 3 | 451 | 70 | ||
NM_079149.5 | NP_523873.2 | 17 - 18 | 6449 | 70 | ||
GenBank mRNA | BT010042.1 | - | 16 - 17 | 5388 | 70 | |
BT120378.1 | - | 2 - 3 | 758 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |