Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103997.3 | NP_001097467.1 | 3 - 4 | 861 | 60 | |
NM_001103998.3 | NP_001097468.1 | 3 - 4 | 861 | 60 | ||
NM_001103999.4 | NP_001097469.2 | 5 - 6 | 1165 | 60 | ||
NM_001104000.2 | NP_001097470.2 | 3 - 4 | 861 | 60 | ||
NM_001274301.1 | NP_001261230.1 | 3 - 4 | 861 | 60 | ||
NM_001274302.1 | NP_001261231.1 | 3 - 4 | 861 | 60 | ||
NM_001274303.1 | NP_001261232.1 | 6 - 7 | 1221 | 60 | ||
NM_001274304.1 | NP_001261233.1 | 11 - 12 | 1952 | 60 | ||
NM_001274305.1 | NP_001261234.1 | 11 - 12 | 2533 | 60 | ||
NM_001274307.1 | NP_001261236.1 | 8 - 9 | 1607 | 60 | ||
NM_001274308.1 | NP_001261237.1 | 8 - 9 | 1607 | 60 | ||
NM_001274309.1 | NP_001261238.1 | 8 - 9 | 1607 | 60 | ||
NM_001299966.1 | NP_001286895.1 | 3 - 4 | 861 | 60 | ||
NM_001299967.1 | NP_001286896.1 | 3 - 4 | 861 | 60 | ||
NM_001316474.1 | NP_001303403.1 | 3 - 4 | 861 | 60 | ||
NM_079154.3 | NP_523878.1 | 3 - 4 | 861 | 60 | ||
NM_138229.2 | NP_612073.2 | 3 - 4 | 861 | 60 | ||
NM_167845.3 | NP_728557.1 | 3 - 4 | 733 | 60 | ||
NM_167846.3 | NP_728558.5 | 10 - 11 | 1799 | 60 | ||
GenBank mRNA | AF064703.2 | - | 3 - 4 | 859 | 60 | |
BT011383.1 | - | 3 - 4 | 829 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster Inventoried |