Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dm01821912_g1
See other GLUT1 GE Assays ›
Gene Symbol:
Glut1
Gene Name:
Glucose transporter 1
Gene Aliases:
0074/12, CG1086, CG13908, CG43946, DmGLUT1, DmGluT1, Dmel\CG43946, Dmel_CG43946, Dmglut1, GLU, GLUT1, Glt1, Glu T1, Glut-1, glut1, l(3)61Ea, l(3)L1H, l(3)S007412
Chromosome Location:
Chr.3L: 914074 - 993788 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0264574
Assay ID
Dm01821912_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Glut1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001103997.3
NP_001097467.1
4 - 5
984
107
NM_001103998.3
NP_001097468.1
4 - 5
984
107
NM_001103999.4
NP_001097469.2
6 - 7
1288
107
NM_001104000.2
NP_001097470.2
4 - 5
984
107
NM_001274301.1
NP_001261230.1
4 - 5
984
107
NM_001274302.1
NP_001261231.1
4 - 5
984
107
NM_001274303.1
NP_001261232.1
7 - 8
1344
107
NM_001274304.1
NP_001261233.1
12 - 13
2075
107
NM_001274305.1
NP_001261234.1
12 - 13
2656
107
NM_001274307.1
NP_001261236.1
9 - 10
1730
107
NM_001274308.1
NP_001261237.1
9 - 10
1730
107
NM_001274309.1
NP_001261238.1
9 - 10
1730
107
NM_001299966.1
NP_001286895.1
4 - 5
984
107
NM_001299967.1
NP_001286896.1
4 - 5
984
107
NM_001316474.1
NP_001303403.1
4 - 5
984
107
NM_079154.3
NP_523878.1
4 - 5
984
107
NM_138229.2
NP_612073.2
4 - 5
984
107
NM_167845.3
NP_728557.1
4 - 5
856
107
NM_167846.3
NP_728558.5
11 - 12
1922
107
GenBank mRNA
AF064703.2
-
4 - 5
982
107
BT011383.1
-
4 - 5
952
107
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
Drosophila melanogaster
Gene Ontology Categories:
Function(s)
Process(es)
glucose homeostasis
transmembrane transport
positive regulation of peptide hormone secretion
transmembrane transporter activity
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0