Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RpL23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001370049.1 | NP_001356972.1 | 2 - 3 | 471 | 80 | |
NM_079162.4 | NP_523886.1 | 2 - 3 | 471 | 80 | ||
GenBank mRNA | BT030798.1 | - | 2 - 3 | 495 | 80 | |
KJ808685.1 | - | 2 - 3 | 485 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |