Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mu2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169855.2 | NP_001163326.1 | 7 - 8 | 2008 | 78 | |
NM_079163.4 | NP_523887.2 | 7 - 8 | 2026 | 78 | ||
NM_167927.3 | NP_728695.1 | 7 - 8 | 1963 | 78 | ||
GenBank mRNA | AY061475.1 | - | 7 - 8 | 2064 | 78 | |
BT030128.1 | - | 7 - 8 | 1987 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |