Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Or63a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169860.1 | NP_001163331.1 | 2 - 3 | 442 | 118 | |
NM_079171.4 | NP_523895.2 | 2 - 3 | 442 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Drosophila melanogaster |