Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eip63E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043116.3 | NP_001036581.1 | 2 - 3 | 1152 | 74 | |
NM_001043117.3 | NP_001036582.1 | 2 - 3 | 202 | 74 | ||
NM_001043118.3 | NP_001036583.1 | 3 - 4 | 507 | 74 | ||
NM_001043119.3 | NP_001036584.1 | 4 - 5 | 316 | 74 | ||
NM_001043120.3 | NP_001036585.1 | 3 - 4 | 535 | 74 | ||
NM_001043121.2 | NP_001036586.1 | 2 - 3 | 413 | 74 | ||
NM_001169869.2 | NP_001163340.1 | 2 - 3 | 888 | 74 | ||
NM_001202113.2 | NP_001189042.1 | 3 - 4 | 558 | 74 | ||
NM_001274444.1 | NP_001261373.1 | 2 - 3 | 1152 | 74 | ||
NM_079180.4 | NP_523904.1 | 2 - 3 | 1152 | 74 | ||
NM_168033.3 | NP_728888.1 | 3 - 4 | 558 | 74 | ||
NM_168034.3 | NP_728889.1 | 4 - 5 | 316 | 74 | ||
NM_168035.2 | NP_728890.1 | 3 - 4 | 535 | 74 | ||
NM_168036.3 | NP_728891.1 | 3 - 4 | 507 | 74 | ||
GenBank mRNA | AF152398.1 | - | 2 - 3 | 1217 | 74 | |
AF152399.1 | - | 2 - 3 | 1217 | 74 | ||
AF152400.1 | - | 3 - 4 | 626 | 74 | ||
AF152401.1 | - | 3 - 4 | 533 | 74 | ||
AF152402.1 | - | 3 - 4 | 533 | 74 | ||
AF152403.1 | - | 2 - 3 | 202 | 74 | ||
AF152404.1 | - | 4 - 5 | 316 | 74 | ||
AF152405.1 | - | 2 - 3 | 907 | 74 | ||
AF152406.1 | - | 3 - 4 | 503 | 74 | ||
BT010064.1 | - | 2 - 3 | 413 | 74 | ||
BT022123.1 | - | 507 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |