Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scsalpha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202115.2 | NP_001189044.1 | 1 - 2 | 149 | 68 | |
NM_079181.4 | NP_523905.2 | 1 - 2 | 149 | 68 | ||
GenBank mRNA | AY089549.1 | - | 1 - 2 | 131 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |