Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gr63a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144411.1 | NP_001137883.1 | 2 - 3 | 363 | 71 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Recommended Assay Drosophila melanogaster |