Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mge | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079186.4 | NP_523910.1 | 2 - 3 | 413 | 51 | |
NM_168044.1 | NP_728910.1 | 2 - 3 | 216 | 51 | ||
GenBank mRNA | AF241222.1 | - | 2 - 3 | 391 | 51 | |
AY122211.1 | - | 2 - 3 | 391 | 51 | ||
BT089014.1 | - | 2 - 3 | 151 | 51 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |