Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scrt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274461.1 | NP_001261390.1 | 1 - 2 | 1601 | 53 | |
NM_079187.5 | NP_523911.2 | 1 - 2 | 1601 | 53 | ||
GenBank mRNA | BT025207.1 | - | 1 - 2 | 1581 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |