Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gr64c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259681.1 | NP_001246610.1 | 9 - 10 | 2386 | 113 | |
NM_079189.4 | NP_523913.2 | 3 - 4 | 1070 | 113 | ||
GenBank mRNA | BT132939.1 | - | 3 - 4 | 1070 | 113 |
Gene Symbol: Gr64b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_168049.2 | NP_728921.1 | 9 - 10 | 2386 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |