Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sse | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079211.3 | NP_523935.1 | 4 - 5 | 1196 | 70 | |
GenBank mRNA | AY119579.1 | - | 4 - 5 | 1134 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |