Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079214.4 | NP_523938.2 | 1 - 2 | 534 | 97 | |
GenBank mRNA | AY089550.1 | - | 3420 | 97 | ||
BT089013.1 | - | 1 - 2 | 391 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |