Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ets65A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144422.2 | NP_001137894.2 | 6 - 7 | 1804 | 60 | |
NM_001316444.1 | NP_001303373.1 | 6 - 7 | 1804 | 60 | ||
NM_001316445.1 | NP_001303374.1 | 6 - 7 | 1804 | 60 | ||
NM_079221.3 | NP_523945.2 | 3 - 4 | 842 | 60 | ||
NM_168156.2 | NP_729142.1 | 6 - 7 | 1804 | 60 | ||
GenBank mRNA | AY060316.1 | - | 3 - 4 | 825 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |