Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079223.2 | NP_523947.2 | 5 - 6 | 3807 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |