Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079223.2 | NP_523947.2 | 7 - 8 | 5421 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |