Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079225.3 | NP_523949.2 | 2 - 3 | 406 | 87 | |
GenBank mRNA | AF367369.1 | - | 2 - 3 | 392 | 87 | |
AY095010.1 | - | 2 - 3 | 385 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |