Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: corn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274567.1 | NP_001261496.1 | 4 - 5 | 3645 | 62 | |
NM_001274568.1 | NP_001261497.1 | 4 - 5 | 3645 | 62 | ||
NM_079230.3 | NP_523954.2 | 4 - 5 | 3645 | 62 | ||
NM_168193.3 | NP_729220.2 | 4 - 5 | 3645 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |