Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: qm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169899.2 | NP_001163370.1 | 5 - 6 | 1024 | 80 | |
NM_001274581.1 | NP_001261510.1 | 5 - 6 | 988 | 80 | ||
NM_079234.4 | NP_523958.2 | 5 - 6 | 1024 | 80 | ||
GenBank mRNA | AY071599.1 | - | 5 - 6 | 990 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |