Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274686.1 | NP_001261615.1 | 5 - 6 | 2184 | 108 | |
NM_001274687.1 | NP_001261616.1 | 5 - 6 | 2184 | 108 | ||
NM_001274688.1 | NP_001261617.1 | 5 - 6 | 2184 | 108 | ||
NM_079267.4 | NP_523991.2 | 4 - 5 | 844 | 108 | ||
NM_168321.3 | NP_729461.1 | 5 - 6 | 2184 | 108 | ||
NM_168322.3 | NP_729462.2 | 5 - 6 | 2280 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |