Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RpS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300079.1 | NP_001287008.1 | 2 - 3 | 441 | 72 | |
NM_079280.4 | NP_524004.2 | 3 - 4 | 469 | 72 | ||
NM_168350.2 | NP_729506.1 | 2 - 3 | 441 | 72 | ||
GenBank mRNA | BT088955.1 | - | 2 - 3 | 443 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |