Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ilp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079288.3 | NP_524012.1 | 1 - 2 | 234 | 77 | |
GenBank mRNA | AJ291726.1 | - | 1 - 2 | 233 | 77 | |
AY069095.1 | - | 1 - 2 | 234 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster Inventoried |