Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mRpL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079298.4 | NP_524022.1 | 1 - 2 | 559 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |