Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TfIIEalpha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079302.4 | NP_524026.1 | 2 - 3 | 1292 | 76 | |
GenBank mRNA | AY119499.1 | - | 2 - 3 | 1284 | 76 | |
U72892.1 | - | 2 - 3 | 1253 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |