Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrx-IV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079310.3 | NP_524034.2 | 9 - 10 | 3517 | 89 | |
NM_168491.3 | NP_729787.2 | 9 - 10 | 3517 | 89 | ||
GenBank mRNA | BT120004.1 | - | 9 - 10 | 3484 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |