Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eyg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014582.2 | NP_001014582.1 | 3 - 4 | 790 | 95 | |
NM_001300118.1 | NP_001287047.1 | 2 - 3 | 857 | 95 | ||
NM_079318.3 | NP_524042.2 | 2 - 3 | 857 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |