Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptp69D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079324.3 | NP_524048.2 | 8 - 9 | 3892 | 91 | |
NM_176324.2 | NP_788502.1 | 8 - 9 | 3889 | 91 | ||
GenBank mRNA | BT001531.1 | - | 8 - 9 | 3885 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |