Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nuf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038920.2 | NP_001034009.1 | 2 - 3 | 876 | 59 | |
NM_001038922.2 | NP_001034011.1 | 2 - 3 | 876 | 59 | ||
NM_001259820.2 | NP_001246749.1 | 2 - 3 | 879 | 59 | ||
NM_001259824.1 | NP_001246753.1 | 2 - 3 | 876 | 59 | ||
GenBank mRNA | BT133122.1 | - | 2 - 3 | 876 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |