Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pka-C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079373.4 | NP_524097.2 | 3 - 4 | 1737 | 88 | |
NM_168638.3 | NP_730083.2 | 4 - 5 | 1055 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |