Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014587.4 | NP_001014587.1 | 1 - 2 | 783 | 63 | |
NM_079382.7 | NP_524106.3 | 1 - 2 | 827 | 63 | ||
GenBank mRNA | AY061352.1 | - | 1 - 2 | 838 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |