Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galphaf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079394.3 | NP_524118.1 | 2 - 3 | 567 | 83 | |
GenBank mRNA | BT064643.1 | - | 2 - 3 | 550 | 83 | |
L09700.1 | - | 2 - 3 | 561 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |