Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Or74a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079399.3 | NP_524123.1 | 1 - 2 | 883 | 76 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Drosophila melanogaster |