Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079413.3 | NP_524137.2 | 1 | 1355 | 139 | |
GenBank mRNA | BT003512.1 | - | 1 | 1353 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |