Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: skl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079415.3 | NP_524139.1 | 1 | 1151 | 172 | |
GenBank mRNA | BT044109.1 | - | 1 | 1146 | 172 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |