Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uggt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079427.3 | NP_524151.2 | 1 - 2 | 435 | 58 | |
GenBank mRNA | BT044158.1 | - | 1 - 2 | 414 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |