Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acp76A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079429.2 | NP_524153.1 | 1 - 2 | 987 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |