Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kap-alpha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079443.2 | NP_524167.1 | 3 - 4 | 799 | 58 | |
GenBank mRNA | AF074957.1 | - | 2 - 3 | 687 | 58 | |
BT083428.1 | - | 3 - 4 | 787 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |