Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mst77F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079464.3 | NP_524188.1 | 1 - 2 | 435 | 81 | |
GenBank mRNA | BT132683.1 | - | 1 - 2 | 376 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |