Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ac78C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259928.2 | NP_001246857.1 | 15 - 16 | 3488 | 64 | |
NM_001275208.1 | NP_001262137.1 | 21 - 22 | 5287 | 64 | ||
NM_001275209.1 | NP_001262138.1 | 22 - 23 | 5498 | 64 | ||
NM_001275210.1 | NP_001262139.1 | 22 - 23 | 5495 | 64 | ||
NM_079470.5 | NP_524194.3 | 22 - 23 | 5498 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |