Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Roe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137042.4 | NP_610886.2 | 2 - 3 | 489 | 73 | |
GenBank mRNA | BT050467.1 | - | 2 - 3 | 484 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |