Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259374.2 | NP_001246303.1 | 21 - 22 | 3372 | 103 | |
NM_001259375.2 | NP_001246304.1 | 19 - 20 | 3661 | 103 | ||
NM_137058.4 | NP_610902.1 | 18 - 19 | 3604 | 103 | ||
GenBank mRNA | AY069830.1 | - | 18 - 19 | 3924 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |