Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137058.4 | NP_610902.1 | 8 - 9 | 2456 | 68 | |
GenBank mRNA | AY069830.1 | - | 8 - 9 | 2776 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |