Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137084.3 | NP_610928.1 | 1 - 2 | 3277 | 112 | |
GenBank mRNA | BT044222.1 | - | 1 - 2 | 3262 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |