Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ih | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038856.4 | NP_001033945.1 | 15 - 16 | 6464 | 62 | |
NM_001038857.3 | NP_001033946.2 | 10 - 11 | 1859 | 62 | ||
NM_001038858.4 | NP_001033947.1 | 13 - 14 | 7176 | 62 | ||
NM_001038859.4 | NP_001033948.1 | 15 - 16 | 6533 | 62 | ||
NM_001144195.3 | NP_001137667.1 | 14 - 15 | 6437 | 62 | ||
NM_001144196.3 | NP_001137668.1 | 15 - 16 | 6518 | 62 | ||
NM_001259389.1 | NP_001246318.1 | 12 - 13 | 1961 | 62 | ||
NM_001259390.2 | NP_001246319.1 | 12 - 13 | 2000 | 62 | ||
NM_001259391.2 | NP_001246320.1 | 12 - 13 | 2002 | 62 | ||
GenBank mRNA | AY928070.1 | - | 15 - 16 | 3925 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster Inventoried |