Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ih | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038859.4 | NP_001033948.1 | 7 - 8 | 5249 | 98 | |
NM_001144195.3 | NP_001137667.1 | 7 - 8 | 5249 | 98 | ||
NM_001144197.3 | NP_001137669.1 | 7 - 8 | 5249 | 98 | ||
NM_001259388.2 | NP_001246317.1 | 5 - 6 | 827 | 98 | ||
GenBank mRNA | AY060342.1 | - | 5 - 6 | 2021 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |